missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17693-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This protein has 9 isoforms produced by alternative splicing with the molecular weight from 114 kDa to 128 kDa. The antibody is specific to the others except the 60 kDa isoform of ABL2.
This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.Spécification
| ABL2 | |
| Polyclonal | |
| Unconjugated | |
| ABL2 | |
| Abelson related gene protein, ABL2, ABLL, ARG, Tyrosine protein kinase ABL2, Tyrosine protein kinase ARG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11352, 27, 304883 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42684, Q4JIM5 | |
| Abl2 | |
| ABL2 Fusion Protein Ag11860 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu