missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51007-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function.Spécification
ABCB6 | |
Polyclonal | |
Unconjugated | |
ABCB6 | |
ABC, ABC14, ABCB6, EST45597, Mt ABC transporter 3, MTABC3, P glycoprotein related protein, PRP, umat | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10058 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NP58 | |
ABCB6 | |
ABCB6 Fusion Protein Ag0461 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |