Learn More
Porvair Sciences 96 well Polypropylene, 800uL Long dripHydrophilic PVDF, mean pore size 0.45µm, 25/Cs,
Marque:  Porvair Sciences 360023
Encompass_Preferred
96 well Polypropylene, 800uL Long dripHydrophilic PVDF, mean pore size 0.45µm, 25/Cs,
Code produit. 12405847
  • 882.98€ / 25 pièces
Expédition estimée: 22-01-2025