missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ 7950 Series Thermal Transfer Printer Ribbon, 83 mm Width
Thermal transfer ribbon with high resistance against smearing and chemicals.
Marque: Brady™ 236008
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Strong environmental resistance against smearing and chemicals.
- Reliable, sharp-edge print quality maximizes bar code readability
Spécification
Noir | |
Wax, Resin | |
3.3 in. | |
83 mm | |
25.4 mm | |
2011/65/UE et 2015/863 |
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer | |
1050 ft. | |
320 m | |
1 in | |
Open-core Ribbon |