Learn More
Gilac™ 75 L Large Volume Hemispheric Container
75 L Large Volume Hemispheric Container is extremely durable, resistant to chemical products, tough and very strong.
Marque:  Gilac™ G146021
Code nomenclature Nacres: HA.11
Informations supplémentaires : Poids : 3.80000kg
Code produit. 17908721
  • 216.13€ / 1 pièce
Expédition estimée: 18-09-2024
pour voir le stock disponible 
Description
Description
  • Large volume hemispheric container.
  • Extremely durable.
  • Resistance to chemical products.
  • Tough and very strong.
  • Volume in litres is equivalent to the container filled to the brim.
Spécification
Spécification
Storage Tote
White
19.8 gal.
HDPE
Semi-circular
75 L