Learn More
Brady™ 6100 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with outstanding durability and reduced energy requirements.
Marque: Brady™ 035282
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Outstanding durability for a general purpose ribbon
- Reduced printhead energy requirements
- 25 mm core
Spécification
Noir | |
Wax, Resin | |
4.3 in. | |
110 mm | |
25.4 mm | |
2011/65/UE et 2015/863 |
BBP72 Label Printer, BBP81 Label Printer, BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer | |
984 ft. | |
300 m | |
1 in | |
Open-core Ribbon |