missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAY 10465, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Description
CAY 10465 is a selective and high-affinity AhR agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM).
Specifications
Specifications
| Nom chimique ou matériau | CAY 10465 |
| Forme physique | Solid |
| CAS | 688348-33-6 |
| Couleur | Blanc |
| Quantité | 50 mg |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C15H9Cl2F3 |
| À utiliser avec (application) | COVID-19-immunoregulation |
| Informations sur la solubilité | DMSO : ≥ 100 mg/mL (315.33 mM) |
| SMILES | FC(C1=CC=C(/C=C/C2=CC(Cl)=CC(Cl)=C2)C=C1)(F)F |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction