Learn More
Brady™ 4300 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with strong resistance against smearing and chemicals.
Marque:  Brady™ 010615
Code produit. 11562535
  • 57.00€ / 1 pièce
En stock
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
Strong environmental resistance against smearing and chemicals.
  • Reliable, sharp-edge print quality maximizes bar code readability
Spécification
Spécification
Noir
Wax, Resin
1 in.
25.4 mm
25.4 mm
2011/65/UE et 2015/863
BBP72 Label Printer, BBP81 Label Printer, BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer
11.8 in.
300 mm
1 in
Open-core Ribbon