Learn More
4-chloro-2-acide fluorocinnamique, 98 %, Thermo Scientific™
Catalog Number 11428876
Click to view available options
Quantité:
5 g
25 g
202982-65-8
C9H5ClFO2
199.59
MFCD00143287
FVLPOWWRHAOKMT-DUXPYHPUSA-M
4-chloro-2-fluorocinnamic acid, 3-4-chloro-2-fluorophenyl acrylic acid, 2e-3-4-chloro-2-fluorophenyl prop-2-enoic acid, 4-chloro-2-fluorocinnamicacid, 3-4-chloro-2-fluorophenyl prop-2-enoic acid, pubchem3431, e-3-4-chloro-2-fluorophenyl acrylic acid, rarechem bk hw 0062, timtec-bb sbb003542, trans-4-chloro-2-fluorocinnamic acid
5941651
(2E)-3-(4-chloro-2-fluorophenyl)prop-2-enoate
[O-]C(=O)\C=C\C1=CC=C(Cl)C=C1F
202982-65-8
C9H5ClFO2
199.59
MFCD00143287
FVLPOWWRHAOKMT-DUXPYHPUSA-M
4-chloro-2-fluorocinnamic acid, 3-4-chloro-2-fluorophenyl acrylic acid, 2e-3-4-chloro-2-fluorophenyl prop-2-enoic acid, 4-chloro-2-fluorocinnamicacid, 3-4-chloro-2-fluorophenyl prop-2-enoic acid, pubchem3431, e-3-4-chloro-2-fluorophenyl acrylic acid, rarechem bk hw 0062, timtec-bb sbb003542, trans-4-chloro-2-fluorocinnamic acid
5941651
(2E)-3-(4-chloro-2-fluorophenyl)prop-2-enoate
[O-]C(=O)\C=C\C1=CC=C(Cl)C=C1F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4-Chloro-2-fluorocinnamic acid
Point de fusion ∼215°C
Quantité 5 g
Poids de la formule 200.6
Pourcentage de pureté 98%