missing translation for 'onlineSavingsMsg'
Learn More

3-(4-bromophényl)-4-(3,4-dichlorophényl)-5-méthyl-4H-1,2,4-triazole, 97 %, Thermo Scientific™

Code produit. 15425966
missing translation for 'orderingAttributeHoverText'
Quantité:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421261-47-3
Formule moléculaire C15H10BrCl2N3
Poids moléculaire (g/mol) 383.07
Numéro MDL MFCD22683144
Clé InChI YHMKSEQFQPFOLL-UHFFFAOYSA-N
Synonyme 3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-4h-1,2,4-triazole
CID PubChem 97030413
Nom IUPAC 3-(4-bromophényl)-4-(3,4-dichlorophényl)-5-méthyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC(=C(C=C2)Cl)Cl)C3=CC=C(C=C3)Br

Specifications

Nom chimique ou matériau 3-(4-Bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-4H-1,2,4-triazole
Quantité 500 mg
Poids de la formule 383.07
Pourcentage de pureté 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.