Learn More
Acide 3-(3-bromo-4-méthoxyphényle)propionique, 96 %, Thermo Scientific™
Code produit. 15475827
Click to view available options
Quantité:
250 mg
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1857-57-4
Formule moléculaire C10H11BrO3
Poids moléculaire (g/mol) 259.10
Numéro MDL MFCD00184737
Clé InChI MLXVZRVWVCOUBQ-UHFFFAOYSA-N
Synonyme 3-3-bromo-4-methoxyphenyl propanoic acid, benzenepropanoicacid, 3-bromo-4-methoxy, 3-3-bromo-4-methoxyphenyl propionic acid, benzenepropanoic acid, 3-bromo-4-methoxy, 3-3-bromo-4-methoxy-phenyl-propionic acid
CID PubChem 4547608
Nom IUPAC Acide 3-(3-bromo-4-méthoxyphényl)propanoïque
SMILES COC1=C(Br)C=C(CCC(O)=O)C=C1
Spécification
Nom chimique ou matériau 3-(3-Bromo-4-methoxyphenyl)propionic acid
Point de fusion 96°C to 100°C
Quantité 1 g
Poids de la formule 259.1
Pourcentage de pureté 96%