missing translation for 'onlineSavingsMsg'
Learn More

Methenolone Enantate, Mikromol™

Code produit. 17929649
Click to view available options
Quantité:
250 mg
Conditionnement:
250mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17929649

Marque: Mikromol MM3721.00

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methenolone Enantate
TRUSTED_SUSTAINABILITY

Spécification

Nom chimique ou matériau Primobolan
CAS 303-42-4
Noms d’analytes ou de composants Primobolan
Conditions d’expédition Room Temperature
Formule moléculaire C27 H42 O3
Certifications/Conformité ISO 17025
Synonyme Metenolone enanthate, Methenolone 17-enanthate, Methenolone enanthate, NSC 64967, Primobolan Depot, Primonabol Depot, SH 601, SQ 16374, (5alpha,17beta)-1-Methyl-17-[(1-oxoheptyl)oxy]-androst-1-en-3-one, 5alpha-Androst-1-en-3-one, 17beta-hydroxy-1-methyl-, heptanoate (6CI,8CI), (5alpha,17beta)-1-Methyl-17-[(1-oxoheptyl)oxy]androst-1-en-3-one, 1-Methyl-17beta-hydroxy-5alpha-androst-1-en-3-one enanthate, 1-Methyl-Delta1-androsten-17beta-ol-3-one enanthate, 17beta-Hydroxy-1-methyl-5alpha-androst-1-en-3-one 17beta-enanthate, 17beta-Hydroxy-1-methyl-5alpha-androst-1-en-3-one heptanoate
Formule InChI InChI=1S/C27H42O3/c1-5-6-7-8-9-25(29)30-24-13-12-22-21-11-10-19-17-20(28)16-18(2)27(19,4)23(21)14-15-26(22,24)3/h16,19,21-24H,5-15,17H2,1-4H3/t19-,21-,22-,23-,24-,26-,27-/m0/s1
SMILES CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C(C)[C@]4(C)[C@H]3CC[C@]12C
Nom IUPAC [(5S,8R,9S,10S,13S,14S,17S)-1,10,13-trimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate
Poids moléculaire (g/mol) 414.62
Quantité 250 mg
Poids de la formule 414.3134 g/mol
Afficher plus Afficher moins
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.