Learn More
2’-désoxyinosine, Thermo Scientific Chemicals
Code produit. 15414019
Click to view available options
Quantité:
1 g
10 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 890-38-0
Formule moléculaire C10H12N4O4
Poids moléculaire (g/mol) 252.23
Numéro MDL MFCD00005762
Clé InChI VGONTNSXDCQUGY-YUZWJPFSNA-N
Synonyme 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
CID PubChem 65058
ChEBI CHEBI:28997
Nom IUPAC 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxyméthyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O
Spécification
Nom chimique ou matériau 2'-Deoxyinosine
Point de fusion >250°C
Couleur Blanc
Quantité 1 g
Beilstein 33517
Poids de la formule 252.23
Forme physique Poudre cristalline