missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-méthylpyrimidine, 99 %, Thermo Scientific Chemicals

Catalog Number p-7030988
Click to view available options
Quantité:
1 g
10 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
13036-57-2
C5H5ClN2
128.559
MFCD00054434
BHAKRVSCGILCEW-UHFFFAOYSA-N
2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride
11629607
2-chloro-4-méthylpyrimidine
CC1=NC(=NC=C1)Cl
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
13036-57-2
C5H5ClN2
128.559
MFCD00054434
BHAKRVSCGILCEW-UHFFFAOYSA-N
2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride
11629607
2-chloro-4-méthylpyrimidine
CC1=NC(=NC=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 2-Chloro-4-methylpyrimidine
Point de fusion 45°C to 48°C
Quantité 1 g
Poids de la formule 128.56
Pourcentage de pureté 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'