Learn More
2,4-Diaminoquinazoline, 98+%, Thermo Scientific Chemicals
Code produit. 15458989
Click to view available options
Quantité:
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1899-48-5
Formule moléculaire C8H8N4
Poids moléculaire (g/mol) 160.18
Numéro MDL MFCD00236032
Clé InChI XELRMPRLCPFTBH-UHFFFAOYSA-N
Synonyme 2,4-diaminoquinazoline, 2,4-quinazolinediamine, quinazolinediamine, fragment 20, acmc-1btzm, 2,4-quinazolinediamine #, 2-aminoquinazolin-4-yl amine, 2,4-diaminoquinazoline, 98+%, 2-amino-quinazolin-4-yl-amine, 2,4-diaminoquinazoline 1g
CID PubChem 65087
Nom IUPAC quinazoline-2,4-diamine
SMILES C1=CC=C2C(=C1)C(=NC(=N2)N)N
Spécification
Nom chimique ou matériau 2,4-Diaminoquinazoline
Point de fusion 254°C to 258°C
Quantité 5 g
Beilstein 131578
Poids de la formule 160.18
Pourcentage de pureté ≥98%