missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1H,1H,2H,3H,3H-perfluoronane-1,2-diol, 95 %
Click to view available options
Quantité:
5g
Spécification
Spécification
Nom chimique ou matériau | 1H,1H,2H,3H,3H-Perfluorononane-1,2-diol |
CAS | 107650-06-6 |
Point d’ébullition | 120°C to 130°C |
Point d’éclair | >150°C (302°F) |
Formule moléculaire | C9H7F13O2 |
Numéro MDL | MFCD00042262 |
Quantité | 5g |
Synonyme | 1h,1h,2h,3h,3h-perfluorononane-1,2-diol, 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluoro-1,2-nonanediol, 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecakis fluoranyl nonane-1,2-diol |
Clé InChI | DAHZCNRVTNHGGR-UHFFFAOYNA-N |
SMILES | OCC(O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'