Learn More
Hamilton™ 1702 SN 25 μL Pre-Calibrated Syringe
Gas tight syringes are ideal for dispensing both liquids and gases. Precision-machined PTFE plunger tip creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample.
Marque:  Hamilton™ CAL80208
Code nomenclature Nacres: SB.52
Informations supplémentaires : Poids : 0.04500kg
Code produit. 17884322
  • 347.00€ / 1 pièce
Expédition estimée: 17-10-2024
pour voir le stock disponible 
Description
Description
Note: This is a Customized item. The following customer specifications are required upon order: Gauge, Length, and Point Style.  
Spécification
Spécification
Not Autoclavable
Custom
25 μL
Gas Sterilizable
Gastight Syringe
1702 SN
Custom (2, 3, 4, 5, or AS)