Learn More
1-[2-(Trifluorométhyl)phényl]pyrrole, 98 %, Thermo Scientific™
Code produit. 15457067
Click to view available options
Quantité:
1 g
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 89108-30-5
Formule moléculaire C11H8F3N
Poids moléculaire (g/mol) 211.187
Numéro MDL MFCD02665223
Clé InChI IUMZRHODYBOXLA-UHFFFAOYSA-N
CID PubChem 11367822
Nom IUPAC 1-[2-(trifluorométhyl)phényl]pyrrole
SMILES C1=CC=C(C(=C1)C(F)(F)F)N2C=CC=C2
Spécification
Nom chimique ou matériau 1-[2-(Trifluoromethyl)phenyl]pyrrole
Point d’ébullition 62°C (1.13 mmHg)
Quantité 1 g
Synonyme 1h-pyrrole, 1-2-trifluoromethyl phenyl, 1-2-trifluoromethyl phenyl pyrrole, 1-2-trifluoromethyl phenyl-1h-pyrrole, acmc-20lhrq, 1-2-trifluoromethyl phenyl-1 h-pyrrole
Poids de la formule 211.19
Pourcentage de pureté 98%