CAS RN 623-58-5
CAS RN 623-58-5
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
Acetoacetic acid sodium salt, MedChemExpress
MedChemExpress Acetoacetic acid sodium salt is a metabolite of non-esterified fatty acids, involved in the development of human diabetes. Acetoacetic acid sodium salt induces oxidative stress to inhibit the assembly of very low density lipoprotein in bovine hepatocytes.