CAS RN 10060-13-6
CAS RN 10060-13-6
IUPAC Name:
diammonium tetrachlorocopperdiuide dihydrate
Synonymes:
diammonium tetrachlorocuprate(2-) dihydrate
Poids moléculaire (g/mol):
277.45
Formule moléculaire:
Cl4CuH12N2O2
InChi Key:
LLMDTWKBBWCWNN-UHFFFAOYSA-L
SMILES:
[NH4+].[NH4+].O.O.Cl[Cu--](Cl)(Cl)Cl
1
–
1
de
1
résultats
1
Cuivre(II) d’ammonium dihydraté, Thermo Scientific Chemicals
CAS: 10060-13-6 Formule moléculaire: Cl4CuH12N2O2 Poids moléculaire (g/mol): 277.454 Numéro MDL: MFCD00150384 Clé InChI: LLMDTWKBBWCWNN-UHFFFAOYSA-L Synonyme: ammonium tetrachlorocuprate ii dihydrate,cupric ammonium chloride dihydrate,ammonium copper ii chloride dihydrate,diammonium tetrachlorocuprate ii dihydrate,ammonium chlorocuprate ii,copper ii ammonium chloride dihydrate,diammonium tetrachlorocuprate dihydrate,copper +2 diammonium chloride dihydrate,diammonium copper ii chloride dihydrate,ammonium tetrachlorocuprated ii dihydrate CID PubChem: 16210985 Nom IUPAC: diazanium ; tétrachlorocuivre(2-) ; dihydrate SMILES: [NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl